Learn More
Sino Biological Monkeypox Virus (MPXV) (Clade Ib) A5L Protein (His Tag)
Catalog No. 502993632
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) A5L Protein (EPI_ISL_19093796) (Met1-Lys281) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3632 Supplier Sino Biological Supplier No. 41048V07E100
A DNA sequence encoding the Monkeypox Virus (MPXV) A5L Protein (EPI_ISL_19093796) (Met1-Lys281) was expressed with a polyhistidine tag at the N-terminus.