Learn More
Sino Biological Monkeypox Virus (MPXV) A5L Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40905T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Monkeypox Virus (MPXV) A5L Protein ( Catalog#40905-V07E). Monkeypox Virus (MPXV) A5L specific IgG was purified by Monkeypox Virus (MPXV) A5L affinity chromatography.
Catalog No. 50-255-3573