Learn More
Sino Biological Monkeypox Virus (MPXV) B6R Antibody, Rabbit MAb
Supplier:  Sino Biological 40902R00420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus (MPXV) B6R (Catalog# 40902-V08H; QNP13760.1; Tyr18-His279).
Catalog No. 50-255-3582