Learn More
Sino Biological Monkeypox Virus (MPXV) B6R Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40902T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Monkeypox Virus (MPXV) B6R (Catalog#40902-V08H). B6R specific IgG was purified by Monkeypox Virus (MPXV) B6R affinity chromatography.
Catalog No. 50-255-3574