Learn More
Aviva Systems Biology Corp MPZL1 Antibody - middle region (ARP46390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPZL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MPZL1. Alternate names: FLJ21047, PZR, PZR1b, PZRa, PZRb, MPZL1bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59317