Learn More
Aviva Systems Biology Corp MPZL3 Antibody - C-terminal region (ARP62685_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62685_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPZL3. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-739