Learn More
Medchemexpress LLC HY-D0090 50mg Medchemexpress, MQAE CAS:162558-52-3 Purity:>98%
SDP
Catalog No. 502031257
Encompass_Preferred
Medchemexpress, HY-D0090 50mg MQAE CAS:162558-52-3 MQAE is a fluorescent indicator that is quenched via collision with chloride, and is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1257 Supplier Medchemexpress LLC Supplier No. HYD009050MG
Medchemexpress, HY-D0090 50mg MQAE CAS:162558-52-3 MQAE is a fluorescent indicator that is quenched via collision with chloride, and is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.