Learn More
TARGETMOL CHEMICALS INC MQAE 200MG
SDP
Catalog No. 502876198
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MQAE, a fluorescent chloride ion indicator, is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells. MQAE is quenched via collision with chloride.
Catalog No. 50-287-6198 Supplier TARGETMOL CHEMICALS INC Supplier No. T3592200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MQAE, a fluorescent chloride ion indicator, is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells. MQAE is quenched via collision with chloride.