Learn More
Abcam MQAE, Fluorescent chloride ion indicator, 200MG
Catalog No. 502705208
Encompass_Preferred
MW 326.19 Da, Purity >97%. Fluorescent chloride ion indicator. Quenches via collision with chloride. Measures chloride transport in liposome membranes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5208 Supplier Abcam Supplier No. AB145418200MG
MW 326.19 Da, Purity >97%. Fluorescent chloride ion indicator. Quenches via collision with chloride. Measures chloride transport in liposome membranes.
The product is subject to the following: Abcam Restricted Use Statement