Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Nitrate Test Method

Catalog No. M1100910022 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5000 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1100910022 5000 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1100910022 Supplier MilliporeSigma™ Supplier No. 1.10091.0022

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

5000 individually sealed colorimetric nitrate test strips

TRUSTED_SUSTAINABILITY

Specifications

Format Individually Sealed
For Use With (Application) Nitrate testing
Quantity 5000 Strips
Test Method Colorimetric
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.