Learn More
Description
- Biologically degradable polyester foil
- Contain special indicator dyes covalently bound to reagent papers
- Rapid on-the-spot determination
- Tests checked and calibrated using certified buffer solutions
- Buffer solutions are traceable to primary reference materials from NIST™ and PTB
- Suited for all media in environmental analysis and in industrial in-process controls
Specifications
Specifications
| pH Range | 5 to 10 |
| For Use With (Application) | Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination |
| Graduations | 11 |
| Quantity | 600 Pack |
| Storage Requirements | 10°C to 25°C |
| Type | Neutralit pH Indicator Strip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
