Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MQuant™ Non-Bleeding pH Indicator Strips
SCI_ED

Catalog No. S07767 Shop All MilliporeSigma Products
Change view
Click to view available options
pH Range:
0 to 6
2 to 9
4 to 7
5 to 10
6.5 to 10
Quantity:
100 Pack
600 Pack
Catalog No. pH Range Quantity
S07767 5 to 10 100 Pack
S07774 6.5 to 10 600 Pack
S07772 4 to 7 600 Pack
S07766 5 to 10 600 Pack
S07764 0 to 6 600 Pack
S07769 2 to 9 100 Pack
6 options
This item is not returnable. View return policy

Catalog No. S07767

Supplier: MilliporeSigma 91263PBURCCR100CAB/DEL

This item is not returnable. View return policy

Since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it.

  • Biologically degradable polyester foil
  • Contain special indicator dyes covalently bound to reagent papers
  • Rapid on-the-spot determination
  • Tests checked and calibrated using certified buffer solutions
  • Buffer solutions are traceable to primary reference materials from NIST™ and PTB
  • Suited for all media in environmental analysis and in industrial in-process controls

TRUSTED_SUSTAINABILITY

Specifications

pH Range 5 to 10
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
Graduations 11
Quantity 100 Pack
Storage Requirements 10°C to 25°C
Type Neutralit pH Indicator Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.