Learn More
Medchemexpress LLC MR 409 | 1445155-39-4 | 99.5% | 3395.91 | 10 MG
SDP
Catalog No. 5000398857
Encompass_Preferred
MR 409 is a selected growth hormone-releasing hormone (GHRH) agonist known for its neuroprotective effects and ability to inhibit lung cancer growth. It enhances endogenous neurogenesis in cerebral ischemic mice and modulates various cellular pathways.
  • Exhibits neuroprotective effects
  • Inhibits lung cancer growth
  • Suppresses p53 expression in BPAECs
  • Induces activation of JAK2, STAT3, and ERK1/2
  • Decreases LPS-induced PGE2 and 8-iso-PGF2α levels
  • Reduces nociceptive response in mice
Catalog No. 50-003-98857 Supplier Medchemexpress LLC Supplier No. HYP330410MG
MR 409 is a selected growth hormone-releasing hormone (GHRH) agonist known for its neuroprotective effects and ability to inhibit lung cancer growth. It enhances endogenous neurogenesis in cerebral ischemic mice and modulates various cellular pathways.
  • Exhibits neuroprotective effects
  • Inhibits lung cancer growth
  • Suppresses p53 expression in BPAECs
  • Induces activation of JAK2, STAT3, and ERK1/2
  • Decreases LPS-induced PGE2 and 8-iso-PGF2α levels
  • Reduces nociceptive response in mice