Learn More
Alkali Scientific MR VP Broth 2mL 13x100mm Screw Cap Polycarbonate, 10/Pk
SDP
Catalog No. 5000663151
Encompass_Preferred
MR VP Broth, 2mL, 13x100mm Screw Cap Polycarbonate Tubes, 10/Pk is a differential culture medium used for biochemical identification of enteric bacteria based on their glucose fermentation pathways. The medium supports both the Methyl Red (MR) test, which detects stable acid production, and the Voges Proskauer (VP) test, which identifies acetoin formation as a neutral fermentation byproduct. These complementary reactions are commonly used as part of IMViC testing for differentiating members of the Enterobacteriaceae family. Each sterile tube contains 2mL of prepared broth in durable screw cap polycarbonate tubes that ensure safe handling and contamination control. This medium is routinely used in clinical microbiology, food safety testing, and environmental microbial identification workflows.
Catalog No. 50-006-63151 Supplier Alkali Scientific Supplier No. MRVPT00
MR VP Broth, 2mL, 13x100mm Screw Cap Polycarbonate Tubes, 10/Pk is a differential culture medium used for biochemical identification of enteric bacteria based on their glucose fermentation pathways. The medium supports both the Methyl Red (MR) test, which detects stable acid production, and the Voges Proskauer (VP) test, which identifies acetoin formation as a neutral fermentation byproduct. These complementary reactions are commonly used as part of IMViC testing for differentiating members of the Enterobacteriaceae family. Each sterile tube contains 2mL of prepared broth in durable screw cap polycarbonate tubes that ensure safe handling and contamination control. This medium is routinely used in clinical microbiology, food safety testing, and environmental microbial identification workflows.