Learn More
Cayman Chemical MR1 ImunoaFInIty Sorbent 400uL
SDP
Catalog No. 502916441
Encompass_Preferred
An inhibitor of MKNK1 and MKNK2 (IC50s 0.064 and 0.086 UM respectively) selective for MKNK1 and MKNK2 over a panel of 104 kinases at 1 UM inhibits phosphorylation of eIF4E in HeLa cells (IC50 0.321 UM) enhances dasatinib-induced tumor growth inhibition in a K562 mouse xenograft model prevents weight gain in a mouse model of high-fat diet-induced obesity at 100 mg/kg
Catalog No. 50-291-6441 Supplier Cayman Chemical Supplier No. 38215400 UL
An inhibitor of MKNK1 and MKNK2 (IC50s 0.064 and 0.086 UM respectively) selective for MKNK1 and MKNK2 over a panel of 104 kinases at 1 UM inhibits phosphorylation of eIF4E in HeLa cells (IC50 0.321 UM) enhances dasatinib-induced tumor growth inhibition in a K562 mouse xenograft model prevents weight gain in a mouse model of high-fat diet-induced obesity at 100 mg/kg