Learn More
Cayman Chemical MR12MIcroglobulIn Complex 20ug
SDP
Catalog No. 502991298
Encompass_Preferred
Source: Recombinant human MR1 and β2-microglobulin expressed in E. coli • Amino acids: 22-292 (MR1), 21-119 (β2-microglobulin) • MW: 32 (MR1) and 12 (β2-microglobulin) kDa
Catalog No. 50-299-1298 Supplier Cayman Chemical Supplier No. 3058520 UG
Source: Recombinant human MR1 and β2-microglobulin expressed in E. coli • Amino acids: 22-292 (MR1), 21-119 (β2-microglobulin) • MW: 32 (MR1) and 12 (β2-microglobulin) kDa