Learn More
Hach Company Hach Lamp, Source, Constant Color - Replacement MR16 Tungsten-Halogen Lamp for QC8000 and QC8500 Analyzers
Catalog No. NC2011744 Shop All Hach Company Products
Encompass
The Hach Lamp, Source, Constant Color (MPN 31069) is a replacement MR16 tungsten-halogen lamp engineered to provide a stable, constant-color light source for analytical instruments such as QC8000 and QC8500 series analyzers. It offers focused illumination and extended service life for routine instrument maintenance.
  • Provides consistent constant-color output for stable measurements.
  • 50 W, 12 V MR16 lamp with GX5.3/GU5.3 two-pin base.
  • Color temperature approximately 3050 K for warm, constant illumination.
  • Approximately 895 lumens output and rated life around 4,000 hours.
  • Compact 1.75 in overall length for direct fit in MR16 lamp holders.
  • Suitable for routine instrument maintenance and straightforward replacement.
Catalog No. NC2011744 Supplier Hach Company Supplier No. 31069
The Hach Lamp, Source, Constant Color (MPN 31069) is a replacement MR16 tungsten-halogen lamp engineered to provide a stable, constant-color light source for analytical instruments such as QC8000 and QC8500 series analyzers. It offers focused illumination and extended service life for routine instrument maintenance.
  • Provides consistent constant-color output for stable measurements.
  • 50 W, 12 V MR16 lamp with GX5.3/GU5.3 two-pin base.
  • Color temperature approximately 3050 K for warm, constant illumination.
  • Approximately 895 lumens output and rated life around 4,000 hours.
  • Compact 1.75 in overall length for direct fit in MR16 lamp holders.
  • Suitable for routine instrument maintenance and straightforward replacement.