Learn More
Cary Company MR50-5 Rieke Pail Pump
SDP
Catalog No. NC1641151
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
26WRPP is a Rieke MR50-5 Pail Pump that comes with an adapter for use on 5 gallon pails with either Rieke Flexspouts or 3/4" and 2" threaded drum openings. Please note if using the MR50-5 on a Rieke Flexspout II, an additional adapter is required. The MR50-5 Rieke Pail Pump dispenses 8 ounces per stroke and can be adjusted to dispense 2 oz, 4 oz, or 6 fl. oz. with special restrictor clips that are included with each pump.
Catalog No. NC1641151 Supplier Cary Company Supplier No. 26WRPP
This item has a minimum qty of 5 per supplier requirements.
26WRPP is a Rieke MR50-5 Pail Pump that comes with an adapter for use on 5 gallon pails with either Rieke Flexspouts or 3/4" and 2" threaded drum openings. Please note if using the MR50-5 on a Rieke Flexspout II, an additional adapter is required. The MR50-5 Rieke Pail Pump dispenses 8 ounces per stroke and can be adjusted to dispense 2 oz, 4 oz, or 6 fl. oz. with special restrictor clips that are included with each pump.