Learn More
Alkali Scientific MR9600-T SmartReader 96 Microp
SDP
Catalog No. 503029528
Encompass_Preferred
For ELISA Testing Intuitive 7 inch color touch screen control panel Filter based system with wavelength range from 340 to 750nm Operates as a stand alone system with USB flash drive drive for data transfer Absorbance range 0 0 to 4 000 Abs 8 channel vertical optical path with zero dispersion Integrated multi-speed plate shaker Temperature controlled plate incubation The SmartReader 96 comes complete with 4 popular filters 405450492630nm and twenty six additional filters are available separately to cover a full range from 340 to 750nm The integrated filter wheel allows up to 8 filters to be loaded at once and filters can be selected during programming Use the touch screen or connect a mouse for programming and operation The menu interface allows the user to choose the wavelength filter set shaking parameters enter details of the plate layout and also choose the details for the reading protocol and data calculations Model MR9600-T offers precise temperature control for incubation of
Catalog No. 50-302-9528 Supplier Alkali Scientific Supplier No. RS7541
Add to Cart
Add to Cart
Request a Quote
For ELISA Testing Intuitive 7 inch color touch screen control panel Filter based system with wavelength range from 340 to 750nm Operates as a stand alone system with USB flash drive drive for data transfer Absorbance range 0 0 to 4 000 Abs 8 channel vertical optical path with zero dispersion Integrated multi-speed plate shaker Temperature controlled plate incubation The SmartReader 96 comes complete with 4 popular filters 405450492630nm and twenty six additional filters are available separately to cover a full range from 340 to 750nm The integrated filter wheel allows up to 8 filters to be loaded at once and filters can be selected during programming Use the touch screen or connect a mouse for programming and operation The menu interface allows the user to choose the wavelength filter set shaking parameters enter details of the plate layout and also choose the details for the reading protocol and data calculations Model MR9600-T offers precise temperature control for incubation of