Learn More
Alkali Scientific Alkali Scientific MR9610 SmartReader Microplate Reader UV-Vis
SDP
Catalog No. 503029634
Encompass_Preferred
The Alkali Scientific MR9610 SmartReader™ UV-Vis Microplate Readers (models MR9610 and MR9611) have been designed with ease-of-use and fast results in mind. These instruments brings extensive capabilities to the lab bench and are designed for the busy lab with multiple users. A large touch screen command center is extremely user-friendly and allows for fast set up of new protocols, as well as modifying and running existing protocols. Intuitive, 10-inch color touch screen control panel Accepts both 96 & 384-well plates Monochromator-based system• No filters required• Wavelength range from 200 to 1000 nm Stand-alone design• No PC or special software required• USB port for flash drive data transfer
Catalog No. 50-302-9634 Supplier Alkali Scientific Supplier No. MR9610
Add to Cart
Add to Cart
Request a Quote
The Alkali Scientific MR9610 SmartReader™ UV-Vis Microplate Readers (models MR9610 and MR9611) have been designed with ease-of-use and fast results in mind. These instruments brings extensive capabilities to the lab bench and are designed for the busy lab with multiple users. A large touch screen command center is extremely user-friendly and allows for fast set up of new protocols, as well as modifying and running existing protocols. Intuitive, 10-inch color touch screen control panel Accepts both 96 & 384-well plates Monochromator-based system• No filters required• Wavelength range from 200 to 1000 nm Stand-alone design• No PC or special software required• USB port for flash drive data transfer