Learn More
Alkali Scientific MR9620 SmartReader MultiMode P
SDP
Catalog No. 503029512
Encompass_Preferred
Expand your discovery horizon and take your research to the next level Intuitive 10-inch color touch screen Stand-alone system with optional PC software (FDA CFR Part 11) Monochromator-based system for UV through visible absorbance Filter module system for fluorescence assays Advanced PMT detector for luminescence assays Optional automatic injection modules for flash luminescence The Alkali Scientific SmartReaderMR9620 Multimode is an advanced microplate reader designed to meet the growing needs of advanced life science labs Equipped with a full suite of high-performance optical systems the SmartReader Multimode is your all-in-one solution for absorbance fluorescence and luminescence applications The SmartReader Multimode quickly processes assays in standard 96 well plates and can also accept 6 12 24 48 and 384 well plates Programming and operation are managed from the instruments large color touch screen and no external computer or software is required Optional PC software is ava
Catalog No. 50-302-9512 Supplier Alkali Scientific Supplier No. MRS9620
Add to Cart
Add to Cart
Request a Quote
Expand your discovery horizon and take your research to the next level Intuitive 10-inch color touch screen Stand-alone system with optional PC software (FDA CFR Part 11) Monochromator-based system for UV through visible absorbance Filter module system for fluorescence assays Advanced PMT detector for luminescence assays Optional automatic injection modules for flash luminescence The Alkali Scientific SmartReaderMR9620 Multimode is an advanced microplate reader designed to meet the growing needs of advanced life science labs Equipped with a full suite of high-performance optical systems the SmartReader Multimode is your all-in-one solution for absorbance fluorescence and luminescence applications The SmartReader Multimode quickly processes assays in standard 96 well plates and can also accept 6 12 24 48 and 384 well plates Programming and operation are managed from the instruments large color touch screen and no external computer or software is required Optional PC software is ava