Learn More
Biomatik Corporation Mras Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11900
Encompass_Preferred
Mras Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MRAS. The Mras Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related Protein M-Ras Protein (79-208Aa). Applications: ELISA, WB, IHC. Target Synonyms: MRAS. Accession Number: O14807. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7555