Learn More
Research Products International Corp Murashige & Skoog MS Medium with Vitamins, 22 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 50213427
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962).
  • Concentration: 4405.1 mg/L
  • Hygroscopic
Catalog No. 50-213-427 Supplier Research Products International Corp Supplier No. M104005.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962).
  • Concentration: 4405.1 mg/L
  • Hygroscopic