Learn More
Research Products International Corp Murashige & Skoog MS Medium with Vitamins, 110 Grams of Powder, Makes 25 Liters of Solution
SDP
Catalog No. 50213426
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962).
  • Concentration: 4405.1 mg/L
  • Hygroscopic
Catalog No. 50-213-426 Supplier Research Products International Corp Supplier No. M1040025.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962).
  • Concentration: 4405.1 mg/L
  • Hygroscopic