Learn More
Research Products International Corp Murashige & Skoog MS Medium, 4.3 Grams of Powder, Makes 1 Liter of Solution
SDP
Catalog No. 50213419
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Murashige and Skoog (1962).
  • Concentration: 4302.09 mg/ml
  • Hygroscopic
Catalog No. 50-213-419 Supplier Research Products International Corp Supplier No. M102001.0
Basal salt mixture containing micro and macro elements as described by Murashige and Skoog (1962).
  • Concentration: 4302.09 mg/ml
  • Hygroscopic