Learn More
Research Products International Corp Murashige & Skoog MS Medium with Gamborg's B5 Vitamins, 110 Grams of Powder, Makes 25 Liters of Solution
SDP
Catalog No. 50213431
Encompass_Preferred
Basal salt mixture containing micro and macro elements with as vitamins as described by Murashige and Skoog (1962) and vitamins as described by Gamborg et. al. (1966).
  • Concentration: 4414.09 mg/L
  • Hygroscopic
Catalog No. 50-213-431 Supplier Research Products International Corp Supplier No. M1050025.0
Basal salt mixture containing micro and macro elements with as vitamins as described by Murashige and Skoog (1962) and vitamins as described by Gamborg et. al. (1966).
  • Concentration: 4414.09 mg/L
  • Hygroscopic