Learn More
Research Products International Corp Murashige & Skoog MS Medium with Gamborg's B5 Vitamins, 220 Grams of Powder, Makes 50 Liters of Solution
SDP
Catalog No. 50213433
Encompass_Preferred
Basal salt mixture containing micro and macro elements with as vitamins as described by Murashige and Skoog (1962) and vitamins as described by Gamborg et. al. (1966).
  • Concentration: 4414.09 mg/L
  • Hygroscopic
Catalog No. 50-213-433 Supplier Research Products International Corp Supplier No. M1050050.0
Basal salt mixture containing micro and macro elements with as vitamins as described by Murashige and Skoog (1962) and vitamins as described by Gamborg et. al. (1966).
  • Concentration: 4414.09 mg/L
  • Hygroscopic