Learn More
Molecular Dimensions MRC PLATE 96 WELL 2 DROP (10)
SDP
Catalog No. 501038532
Encompass
MRC PLATE 96 WELL 2 DROP (10) UV Crystallization Plate was designed by crystallographers at the MRC in Cambridge. UV transmissible polymer with excellent optical properties. Raised, wide wells make crystal retrieval easy.The wells are conical. 10nL-5ul drops and the reservoirs hold volumes from 50-100 ul.
Catalog No. 50-103-8532 Supplier Molecular Dimensions Supplier No. MD1100U10
MRC PLATE 96 WELL 2 DROP (10) UV Crystallization Plate was designed by crystallographers at the MRC in Cambridge. UV transmissible polymer with excellent optical properties. Raised, wide wells make crystal retrieval easy.The wells are conical. 10nL-5ul drops and the reservoirs hold volumes from 50-100 ul.