Learn More
ATCC MRC-9; Lung Fibroblast; Human (Homo sapiens)
Supplier:  ATCC CCL212
Encompass
MRC-9 is a fibroblast cell line that was isolated in 1974 from the lung of a normal White, 15-week-old, female embryo. This cell line was deposited by JP Jacobs.
Catalog No. 50-238-4906