Learn More
Sino Biological Recombinant Human CDK11A/CDC2L2 Protein (GST Tag)
Catalog No. 502575643
Encompass_Preferred
Recombinant human CDK11A (361-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5643 Supplier Sino Biological Supplier No. C2431G100
Recombinant human CDK11A (361-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.