Learn More
Carnabio Usa Inc MRCKA(CDC42BPA)/5UG/TRUNCATED
Catalog No. 507540524
Encompass
MRCK(CDC42BPA), Truncated protein, Wild type, Amino Acid 1-574, NP_003598.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0524 Supplier Carnabio Usa Inc Supplier No. 011075
MRCK(CDC42BPA), Truncated protein, Wild type, Amino Acid 1-574, NP_003598.2, Expressed in Insect (sf21); 5microg