Learn More
Eurofins Discoverx MRCKβ, active, 250 µg. (14-643M)
Supplier:  Eurofins Discoverx 14643M
Encompass
N-terminal 6xHis-tagged recombinant, human MRCKb amino acids 1-473. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9108