Learn More
Medchemexpress LLC MRE-269-d7 | 1265295-20-2 | 99.6% | C25H22D7N3O3 | 1 MG
SDP
Catalog No. 5000433034
Encompass_Preferred
MRE-269-d7 is a deuterium-labeled form of MRE-269. It functions as an active metabolite of selexipag and acts as a selective IP receptor agonist. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development. Deuteration can also impact the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis.
  • It is an isotope-labeled compound.
  • It is suitable for use in in vivo cell experiments.
  • It is useful for therapeutic drug monitoring.
Catalog No. 50-004-33034 Supplier Medchemexpress LLC Supplier No. HY79593S21MG
MRE-269-d7 is a deuterium-labeled form of MRE-269. It functions as an active metabolite of selexipag and acts as a selective IP receptor agonist. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development. Deuteration can also impact the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis.
  • It is an isotope-labeled compound.
  • It is suitable for use in in vivo cell experiments.
  • It is useful for therapeutic drug monitoring.