Learn More
Medchemexpress LLC MRE-269-d7 | 1265295-20-2 | 99.6% | C25H22D7N3O3 | 5 MG
SDP
Catalog No. 5000433100
Encompass_Preferred
MRE-269-d7 is a deuterium-labeled version of MRE-269, an active metabolite of selexipag that acts as a selective IP receptor agonist. It is intended for research use and can serve as a tracer and an internal standard for quantitative analysis. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled version of MRE-269
  • Active metabolite of selexipag
  • Selective IP receptor agonist
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-33100 Supplier Medchemexpress LLC Supplier No. HY79593S25MG
MRE-269-d7 is a deuterium-labeled version of MRE-269, an active metabolite of selexipag that acts as a selective IP receptor agonist. It is intended for research use and can serve as a tracer and an internal standard for quantitative analysis. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled version of MRE-269
  • Active metabolite of selexipag
  • Selective IP receptor agonist
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs