Learn More
Sino Biological MRE11 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 203501T38100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human MRE11 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-8197