Learn More
Aviva Systems Biology Corp MRE11 Antibody - N-terminal region (ARP56644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRE11A. It was validated on Western Blot. MRE11A,MRE11 meiotic recombination 11 homolog A (S. cerevisiae),ATLD; HNGS1; MRE11; MRE11B; MRE11A,double-strand break repair protein MRE11A,double-strand break repair protein MRE11A,AT-like disease,MRE11 homolog 1,MRE11 homolog A,endo/exonuclease Mre11,meiotic recombination 11 homolog 1,meiotic recombination 11 homolog A,DNA recombination and repair proteinThe immunogen is a synthetic peptide directed towards the N terminal region of human MRE11A. Alternate names: ATLD, HNGS1, MRE11, MRE11BApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-185