Learn More
Cayman Chemical MRE11humnrecombIntaa 100ug
SDP
Catalog No. 502942541
Encompass_Preferred
Source: Recombinant human N-terminal His- and GST-tagged MRE11 expressed in E. coli • Amino acids: 1-411 • MW: 74.7 kDa
Catalog No. 50-294-2541 Supplier Cayman Chemical Supplier No. 40752100 UG
Source: Recombinant human N-terminal His- and GST-tagged MRE11 expressed in E. coli • Amino acids: 1-411 • MW: 74.7 kDa