Learn More
Cayman Chemical MRE11humnrecombIntaa 10ug
SDP
Catalog No. 502942521
Encompass_Preferred
Source: Recombinant human N-terminal His- and GST-tagged MRE11 expressed in E. coli • Amino acids: 1-411 • MW: 74.7 kDa
Catalog No. 50-294-2521 Supplier Cayman Chemical Supplier No. 4075210 UG
Source: Recombinant human N-terminal His- and GST-tagged MRE11 expressed in E. coli • Amino acids: 1-411 • MW: 74.7 kDa