Learn More
Bachem H-Met-Arg-Phe-Ala-OH
SDP
Catalog No. NC9634265
Encompass_Preferred
H-Met-Arg-Phe-Ala-OH, 67368-29-0, C23H37N7O5S, Mr 523.66, 50mg. MRFA. MRFA is used as a calibration standard in mass spectrometry (ESI). Miao et al. studied Pt(II) complexes of the tetrapeptide by mass spectrometric methods. MRFA has been shown to be a competitive inhibitor of an enkephalin-generating endopeptidase isolated from rat brain. The peptide is a substrate for dipeptidyl peptidase III from human erythrocytes and for snapalysin.
Catalog No. NC9634265 Supplier Bachem Supplier No. 4004330.0050
Add to Cart
Add to Cart
H-Met-Arg-Phe-Ala-OH, 67368-29-0, C23H37N7O5S, Mr 523.66, 50mg. MRFA. MRFA is used as a calibration standard in mass spectrometry (ESI). Miao et al. studied Pt(II) complexes of the tetrapeptide by mass spectrometric methods. MRFA has been shown to be a competitive inhibitor of an enkephalin-generating endopeptidase isolated from rat brain. The peptide is a substrate for dipeptidyl peptidase III from human erythrocytes and for snapalysin.