Learn More
Eurofins Discoverx ChemiScreen MRGPRD Receptor Membrane Preparation, 1 vial. (HTS206M)
Supplier:  Eurofins Discoverx HTS206M
Encompass
The ChemiScreen MRGPRD Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of MRGPRD surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-6052