Learn More
Eurofins Discoverx ChemiScreen MRGPRD Receptor Stable Cell Line, 2 vials. (HTS206C)
Catalog No. 502006051
Encompass
The ChemiScreen MRGPRD Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. β-alanine) based activation of MRGPRD GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses MRGPRD alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of MRGPRD, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6051 Supplier Eurofins Discoverx Supplier No. HTS206C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen MRGPRD Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. β-alanine) based activation of MRGPRD GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses MRGPRD alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of MRGPRD, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.