Learn More
Aviva Systems Biology Corp MRGPRE Antibody - C-terminal region (ARP62973_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62973_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRGPRE. It was validated on Western Blot. . Alternate names: GPR167, MGC138408, MRGEApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-287-825