Learn More
Aviva Systems Biology Corp MRGPRF Antibody - C-terminal region (ARP59920_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59920_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRGPRF. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MRGPRF. Alternate names: RTA, MRGF, GPR140, GPR168, FLJ16111, FLJ29034, FLJ40998, FLJ53714, MGC21621, DKFZp586B2122, MRGPRF, mrgFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-876