Learn More
Medchemexpress LLC MRGPRX1 agonist 1 | 2377379-39-8 | 99.95% | 100 MG
SDP
Catalog No. 5000339013
Encompass_Preferred
MRGPRX1 agonist 1 is a highly potent agonist for MRGPRX1 with an EC50 of 50 nM. It exhibits over 50-fold selectivity for δ, μ, and κ opioid receptors, showing inactivity against MRGPRC11. This compound is valuable for researching chronic pain, particularly neuropathic pain, by mitigating nociceptive transmission in the spinal cord.
  • Highly potent MRGPRX1 agonist (EC50: 50 nM)
  • Over 50-fold selectivity for δ, μ, and κ opioid receptors
  • Inactive against MRGPRC11
  • Useful for the study of chronic pain, especially neuropathic pain
  • Demonstrates good distribution to the spinal cord in male mice
  • Binding affinities (Kis) for δ-Opioid (2.47 μM), μ-opioid (3.73 μM), and κ-opioid (12.10 μM) receptors
Catalog No. 50-003-39013 Supplier Medchemexpress LLC Supplier No. HY130118100MG
MRGPRX1 agonist 1 is a highly potent agonist for MRGPRX1 with an EC50 of 50 nM. It exhibits over 50-fold selectivity for δ, μ, and κ opioid receptors, showing inactivity against MRGPRC11. This compound is valuable for researching chronic pain, particularly neuropathic pain, by mitigating nociceptive transmission in the spinal cord.
  • Highly potent MRGPRX1 agonist (EC50: 50 nM)
  • Over 50-fold selectivity for δ, μ, and κ opioid receptors
  • Inactive against MRGPRC11
  • Useful for the study of chronic pain, especially neuropathic pain
  • Demonstrates good distribution to the spinal cord in male mice
  • Binding affinities (Kis) for δ-Opioid (2.47 μM), μ-opioid (3.73 μM), and κ-opioid (12.10 μM) receptors