Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MRGPRX1 agonist 1 | 2377379-39-8 | 99.95% | 50 MG
SDP

Catalog No. 5000340985
Encompass_Preferred

MRGPRX1 agonist 1 is a highly potent MRGPRX1 agonist with an EC50 of 50 nM. It exhibits greater than 50-fold selectivity for δ, μ, and κ opioid receptors and is inactive against MRGPRC11. This product is useful for the study of chronic pain, particularly neuropathic pain.

  • Highly potent MRGPRX1 agonist with an EC50 of 50 nM
  • Exhibits greater than 50-fold selectivity for δ, μ, and κ opioid receptors
  • Inactive against MRGPRC11
  • Inhibits high voltage-activated (HVA) Ca2+ currents
  • Reduces neurotransmitter release
  • Mitigates nociceptive transmission in the spinal cord
  • Useful for the study of chronic pain, particularly neuropathic pain
  • Molecular weight of 435.50
  • Formula of C23H21N3O4S
  • Appearance is a white to yellow solid
  • For research use only

Catalog No. 50-003-40985 Supplier Medchemexpress LLC Supplier No. HY13011850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MRGPRX1 agonist 1 is a highly potent MRGPRX1 agonist with an EC50 of 50 nM. It exhibits greater than 50-fold selectivity for δ, μ, and κ opioid receptors and is inactive against MRGPRC11. This product is useful for the study of chronic pain, particularly neuropathic pain.

  • Highly potent MRGPRX1 agonist with an EC50 of 50 nM
  • Exhibits greater than 50-fold selectivity for δ, μ, and κ opioid receptors
  • Inactive against MRGPRC11
  • Inhibits high voltage-activated (HVA) Ca2+ currents
  • Reduces neurotransmitter release
  • Mitigates nociceptive transmission in the spinal cord
  • Useful for the study of chronic pain, particularly neuropathic pain
  • Molecular weight of 435.50
  • Formula of C23H21N3O4S
  • Appearance is a white to yellow solid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.