Learn More
Medchemexpress LLC Mrgprx1 agonist 1 | 2377379-39-8 | C23H21N3O4S | 10MG
SDP
Catalog No. 5000261850
Encompass_Preferred
MRGPRX1 agonist 1 is a potent, selective small-molecule agonist of the Mas-related G protein-coupled receptor X1 (MRGPRX1). It has an EC50 of 50 nM and demonstrates >50-fold selectivity over δ, μ, and κ opioid receptors while showing no activity at MRGPRC11. It is used as a research reagent in studies of nociception and neuropathic pain signaling.
  • Potent MRGPRX1 agonist (EC50 50 nM).
  • Greater than 50-fold selectivity over delta, mu, and kappa opioid receptors.
  • Inactive against MRGPRC11.
  • Molecular formula C23H21N3O4S and molecular weight 435.50.
  • Intended for research use in nociception and pain pathway studies.
Catalog No. 50-002-61850 Supplier Medchemexpress LLC Supplier No. HY13011810MG
MRGPRX1 agonist 1 is a potent, selective small-molecule agonist of the Mas-related G protein-coupled receptor X1 (MRGPRX1). It has an EC50 of 50 nM and demonstrates >50-fold selectivity over δ, μ, and κ opioid receptors while showing no activity at MRGPRC11. It is used as a research reagent in studies of nociception and neuropathic pain signaling.
  • Potent MRGPRX1 agonist (EC50 50 nM).
  • Greater than 50-fold selectivity over delta, mu, and kappa opioid receptors.
  • Inactive against MRGPRC11.
  • Molecular formula C23H21N3O4S and molecular weight 435.50.
  • Intended for research use in nociception and pain pathway studies.