Learn More
Aviva Systems Biology Corp MRGPRX1 Antibody - C-terminal region (ARP59925_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59925_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRGPRX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human MRGPRX1. Alternate names: GPCR, MRGX1, SNSR4ApplicationWB. Species reactivity: Human
Catalog No. 50-286-879