Learn More
Eurofins Discoverx PathHunter MRGPRX2 Bioassay Kit, 2-Plate. (93-0309Y2-00143)
Catalog No. 502000237
Encompass
The PathHunter MRGPRX2 Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay, including cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility, and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0237 Supplier Eurofins Discoverx Supplier No. 930309Y200143
Add to Cart
Add to Cart
The PathHunter MRGPRX2 Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay, including cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility, and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.