Learn More
Medchemexpress LLC Mrgprx2 modulator-1 (example 17) | 2781839-42-5 | 99.0% | 459.39 g/mol | C20H19F6N5O | 10 MG
SDP
Catalog No. 5000285406
Encompass_Preferred
MRGPRX2 modulator-1 (example 17) is a small-molecule modulator of the Mas-related G-protein coupled receptor X2 (MRGPRX2) supplied for preclinical research. It is intended for studies of inflammation, pain signaling, and autoimmune disorders and is characterized by a confirmed molecular formula, molecular weight, and high HPLC purity.
  • Small-molecule modulator of MRGPRX2 suitable for preclinical research.
  • High purity (99.03% by HPLC) for consistent bioactivity.
  • Molecular formula: C20H19F6N5O; molecular weight: 459.39 g/mol.
  • Assigned CAS number 2781839-42-5 for unambiguous identification.
  • Provided as a powdered research reagent; handle per SDS instructions.
Catalog No. 50-002-85406 Supplier Medchemexpress LLC Supplier No. HY16054010MG
MRGPRX2 modulator-1 (example 17) is a small-molecule modulator of the Mas-related G-protein coupled receptor X2 (MRGPRX2) supplied for preclinical research. It is intended for studies of inflammation, pain signaling, and autoimmune disorders and is characterized by a confirmed molecular formula, molecular weight, and high HPLC purity.
  • Small-molecule modulator of MRGPRX2 suitable for preclinical research.
  • High purity (99.03% by HPLC) for consistent bioactivity.
  • Molecular formula: C20H19F6N5O; molecular weight: 459.39 g/mol.
  • Assigned CAS number 2781839-42-5 for unambiguous identification.
  • Provided as a powdered research reagent; handle per SDS instructions.