Learn More
Aviva Systems Biology Corp MRGPRX3 Antibody - C-terminal region (ARP59922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRGPRX3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MRGPRX3. Alternate names: GPCR, MRGX3, SNSR1, SNSR2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-877